Assay Details
Target Gene Details
Entrez Gene ID: | 23508 |
Gene Name: | tetratricopeptide repeat domain 9 |
Gene Aliases: |
TTC9A |
Location: |
Chr.14:70641787-70675360 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTC9 | NM_015351.1 | NP_056166.1 | ||
BC047950.1 | AAH47950.1 | |||
BX407741.2 | ||||
D86980.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |