Assay Details
Target Gene Details
Entrez Gene ID: | 390502 |
Gene Name: | serpin family A member 2 (gene/pseudogene) |
Gene Aliases: |
ARGS, ATR, PIL, SERPINA2P, psiATR |
Location: |
Chr.14:94364318-94366702 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINA2 | NR_110563.1 | 1 | 559 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832862 | Chr.14:94163327 - 94374084 on Build GRCh38 | Gain | SERPINA6 PPP4R4 SERPINA10 SERPINA2 |
nsv1120301 | Chr.14:94363766 - 94380253 on Build GRCh38 | Tandem Duplication | SERPINA1 SERPINA2 |
nsv509545 | Chr.14:94360156 - 94400412 on Build GRCh38 | Insertion | SERPINA1 SERPINA2 |
nsv832863 | Chr.14:94348482 - 94501487 on Build GRCh38 | Gain | SERPINA9 SERPINA1 SERPINA12 SERPINA2 SERPINA11 |
nsv512370 | Chr.14:94362618 - 94378920 on Build GRCh38 | Loss | SERPINA1 SERPINA2 |
nsv565622 | Chr.14:94342423 - 94375205 on Build GRCh38 | Loss | SERPINA2 |
nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain | SERPINA13P SERPINA6 SERPINA9 SERPINA4 SERPINA1 SERPINA5 SERPINA3 SERPINA12 GSC SERPINA2 SERPINA11 |
esv3635377 | Chr.14:94176964 - 94375000 on Build GRCh38 | Gain | SERPINA6 PPP4R4 SERPINA10 SERPINA2 |
nsv1406 | Chr.14:94361166 - 94394512 on Build GRCh38 | Insertion | SERPINA1 SERPINA2 |
esv2749043 | Chr.14:94361914 - 94370564 on Build GRCh38 | Deletion | SERPINA2 |
nsv832864 | Chr.14:94362101 - 94538150 on Build GRCh38 | Loss | SERPINA9 SERPINA1 SERPINA12 SERPINA2 SERPINA11 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |