Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_001318380.1 | NP_001305309.1 | ||
NM_177533.4 | NP_803877.2 | |||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | ||||
AK310449.1 | ||||
BC041584.1 | AAH41584.1 | |||
DA249638.1 | ||||
DA409636.1 | ||||
DB096906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | MIR6765 NUDT14 JAG2 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | MIR6765 MTA1 BRF1 NUDT14 GPR132 LOC100507437 PACS2 JAG2 LOC102723354 TEX22 BTBD6 CRIP2 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | MTA1 BRF1 NUDT14 C14orf80 PACS2 TEX22 MIR6765 IGH LOC105370697 LOC100507437 TMEM121 CRIP1 JAG2 BTBD6 IGHA2 CRIP2 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | MIR6765 BRF1 NUDT14 GPR132 JAG2 LOC102723354 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | MIR6765 BRF1 NUDT14 JAG2 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | MIR6765 NUDT14 JAG2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | EIF5 ASPG TDRD9 CDCA4 GPR132 LOC107984648 LINC00638 PACS2 LOC105378183 SNORA28 LOC388022 SIVA1 KLC1 MIR203A C14orf180 LINC00637 LOC105370691 ADSSL1 LOC105370687 MARK3 JAG2 CKB PLD4 BTBD6 RD3L BRF1 NUDT14 LOC101929634 ZFYVE21 AHNAK2 CEP170B AKT1 LOC102723354 LOC105370708 BAG5 XRCC3 TRMT61A MIR203B INF2 PPP1R13B MIR6765 APOPT1 ZBTB42 C14orf2 KIF26A TMEM179 MIR4710 C14orf79 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | MIR6765 BRF1 NUDT14 CDCA4 GPR132 AHNAK2 JAG2 LOC102723354 BTBD6 C14orf79 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | ELK2AP IGHG1 IGHV4-55 IGHD PACS2 TEX22 IGHG4 IGH LOC105370697 KIAA0125 LOC100507437 CRIP1 LOC107987211 IGHV3-23 JAG2 MIR4539 BTBD6 IGHV4-31 IGHA2 MIR8071-1 IGHV3-69-1 IGHV3-48 CRIP2 MIR4537 MTA1 IGHV3-30 BRF1 MIR4538 NUDT14 MIR8071-2 C14orf80 IGHV3-41 IGHV1-69 IGHV3-71 MIR6765 IGHA1 MIR4507 TMEM121 LINC00226 IGHG3 IGHG2 ADAM6 LINC00221 IGHM |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | MIR6765 BRF1 NUDT14 CDCA4 GPR132 AHNAK2 JAG2 LOC102723354 C14orf79 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |