Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | CDCA4 GPR132 C14orf79 AHNAK2 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | JAG2 CDCA4 GPR132 LOC102723354 C14orf79 AHNAK2 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | CDCA4 C14orf79 PLD4 AHNAK2 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 PLD4 AHNAK2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | TRMT61A MIR6765 SIVA1 C14orf180 LINC00638 C14orf2 KIF26A PLD4 APOPT1 NUDT14 INF2 CEP170B MIR203A ASPG TDRD9 BTBD6 MIR203B MARK3 EIF5 LOC107984648 KLC1 LOC105370691 PACS2 LOC105370708 TMEM179 AKT1 LOC105370687 CKB LOC105378183 ZFYVE21 BRF1 PPP1R13B LOC102723354 RD3L ADSSL1 AHNAK2 JAG2 CDCA4 ZBTB42 GPR132 LOC101929634 MIR4710 BAG5 C14orf79 XRCC3 SNORA28 LINC00637 LOC388022 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | JAG2 CDCA4 MIR6765 BTBD6 GPR132 BRF1 LOC102723354 C14orf79 AHNAK2 NUDT14 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | JAG2 CDCA4 MIR6765 GPR132 BRF1 LOC102723354 C14orf79 AHNAK2 NUDT14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |