Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | GPR132 C14orf79 AHNAK2 CDCA4 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | GPR132 C14orf79 AHNAK2 JAG2 LOC102723354 CDCA4 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | C14orf79 AHNAK2 PLD4 CDCA4 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 AHNAK2 PLD4 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | CEP170B GPR132 LOC107984648 ADSSL1 BTBD6 RD3L XRCC3 AKT1 PPP1R13B LOC105378183 C14orf2 TRMT61A JAG2 MIR4710 KIF26A NUDT14 MIR6765 KLC1 LOC101929634 EIF5 LOC105370691 ZFYVE21 C14orf79 MARK3 SIVA1 LINC00637 C14orf180 LOC105370687 LINC00638 ASPG PLD4 SNORA28 INF2 PACS2 TDRD9 LOC105370708 APOPT1 AHNAK2 BAG5 TMEM179 CKB LOC102723354 MIR203B ZBTB42 LOC388022 BRF1 MIR203A CDCA4 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | GPR132 C14orf79 BTBD6 AHNAK2 JAG2 LOC102723354 NUDT14 MIR6765 BRF1 CDCA4 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | GPR132 C14orf79 AHNAK2 JAG2 LOC102723354 NUDT14 MIR6765 BRF1 CDCA4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |