Assay Details
Target Gene Details
Entrez Gene ID: | 196913 |
Gene Name: | long intergenic non-protein coding RNA 1599 |
Gene Aliases: |
C14orf183 |
Location: |
Chr.14:50007313-50105043 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01599 | NR_131171.1 | |||
BQ027525.1 | ||||
BU927649.1 | ||||
DA856251.1 | ||||
T90102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1048869 | Chr.14:50020658 - 50056451 on Build GRCh38 | Gain |
![]() |
esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)