Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | C14orf79 AHNAK2 GPR132 CDCA4 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | C14orf79 LOC102723354 AHNAK2 GPR132 JAG2 CDCA4 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | C14orf79 AHNAK2 PLD4 CDCA4 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | C14orf79 AHNAK2 PLD4 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | LINC00638 C14orf79 ZFYVE21 LOC102723354 LINC00637 ADSSL1 PLD4 BTBD6 TMEM179 CKB XRCC3 EIF5 SIVA1 GPR132 JAG2 MIR203A KLC1 AKT1 LOC105370687 PACS2 ASPG LOC105370708 LOC107984648 BRF1 LOC101929634 KIF26A MARK3 LOC105370691 ZBTB42 CDCA4 NUDT14 TRMT61A INF2 SNORA28 CEP170B PPP1R13B AHNAK2 C14orf2 MIR203B MIR6765 RD3L C14orf180 LOC388022 LOC105378183 MIR4710 APOPT1 TDRD9 BAG5 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | C14orf79 LOC102723354 AHNAK2 MIR6765 BRF1 GPR132 JAG2 BTBD6 CDCA4 NUDT14 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | C14orf79 LOC102723354 AHNAK2 MIR6765 BRF1 GPR132 JAG2 CDCA4 NUDT14 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |