Assay Details
Target Gene Details
Entrez Gene ID: | 5891 |
Gene Name: | MOK protein kinase |
Gene Aliases: |
RAGE, RAGE-1, RAGE1, STK30 |
Location: |
Chr.14:102224450-102305200 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOK | NR_073540.1 | |||
XM_006720225.2 | XP_006720288.1 | |||
XM_011537052.1 | XP_011535354.1 | |||
XM_017021555.1 | XP_016877044.1 | |||
XM_017021556.1 | XP_016877045.1 | |||
HY184670.1 | ||||
U46194.1 | AAB38087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv22037 | Chr.14:102223226 - 102248955 on Build GRCh38 | Gain | WDR20 MOK |
nsv1429 | Chr.14:102201072 - 102246584 on Build GRCh38 | Deletion | WDR20 MOK |
esv3635547 | Chr.14:102223491 - 102317911 on Build GRCh38 | Gain | ZNF839 WDR20 MOK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |