Assay Details
Target Gene Details
Entrez Gene ID: | 7517 |
Gene Name: | X-ray repair cross complementing 3 |
Gene Aliases: |
CMM6 |
Location: |
Chr.14:103697611-103715486 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
XRCC3 | NM_001100118.1 | NP_001093588.1 | ||
NM_001100119.1 | NP_001093589.1 | |||
NM_005432.3 | NP_005423.1 | |||
XM_005268046.2 | XP_005268103.1 | |||
XM_011537138.2 | XP_011535440.1 | |||
AF035586.1 | AAC05368.1 | |||
AK022829.1 | ||||
AK023646.1 | ||||
AK124498.1 | ||||
AK126706.1 | ||||
BC001036.1 | AAH01036.1 | |||
BC011725.2 | AAH11725.1 | |||
BT007417.1 | AAP36085.1 | |||
BX161398.1 | CAD61884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | XRCC3 KLC1 LOC107984648 |
esv2749175 | Chr.14:103700180 - 103705264 on Build GRCh38 | Deletion | XRCC3 KLC1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | XRCC3 KLC1 LOC107984648 ZFYVE21 |
nsv952260 | Chr.14:103697564 - 103723263 on Build GRCh38 | Deletion | XRCC3 KLC1 LOC107984648 ZFYVE21 |
nsv527425 | Chr.14:103695926 - 103731914 on Build GRCh38 | Loss | XRCC3 KLC1 LOC107984648 ZFYVE21 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | RD3L GPR132 XRCC3 LOC107984648 AHNAK2 PLD4 MIR203A LOC102723354 ADSSL1 JAG2 CKB AKT1 TRMT61A BTBD6 C14orf2 CDCA4 C14orf180 PPP1R13B SIVA1 EIF5 KIF26A LOC105370708 MIR6765 ZBTB42 APOPT1 BAG5 TMEM179 LOC105378183 LINC00638 MARK3 LOC105370691 MIR203B MIR4710 ASPG KLC1 TDRD9 C14orf79 NUDT14 LINC00637 BRF1 INF2 LOC388022 LOC105370687 ZFYVE21 CEP170B LOC101929634 PACS2 SNORA28 |
esv2760013 | Chr.14:103681188 - 103946476 on Build GRCh38 | Loss | RD3L XRCC3 KLC1 LOC107984648 TDRD9 C14orf2 LINC00637 PPP1R13B ZFYVE21 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |