Assay Details
Target Gene Details
Entrez Gene ID: | 11154 |
Gene Name: | adaptor related protein complex 4 sigma 1 subunit |
Gene Aliases: |
AP47B, CLA20, CLAPS4, CPSQ6, SPG52 |
Location: |
Chr.14:31025106-31096450 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP4S1 | NM_001128126.2 | NP_001121598.1 | ||
NM_001254726.1 | NP_001241655.1 | |||
NM_001254727.1 | NP_001241656.1 | |||
NM_001254728.1 | NP_001241657.1 | |||
NM_001254729.1 | NP_001241658.1 | |||
NM_007077.4 | NP_009008.2 | |||
XM_005267293.4 | XP_005267350.1 | |||
XM_011536371.2 | XP_011534673.1 | |||
XM_011536372.2 | XP_011534674.1 | |||
AB030654.1 | BAA82970.1 | |||
AF155159.1 | AAD43329.1 | |||
AK304115.1 | ||||
AK316517.1 | ||||
BC001259.1 | AAH01259.1 | |||
BF671561.1 | ||||
BG702035.1 | ||||
BX247969.1 | CAD62307.1 | |||
BX388185.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv564169 | Chr.14:30349881 - 31072288 on Build GRCh38 | Gain | MIR624 G2E3 STRN3 AP4S1 LOC100506071 SCFD1 COCH |
nsv832763 | Chr.14:31032391 - 31210815 on Build GRCh38 | Loss | AP4S1 HECTD1 |
nsv1049063 | Chr.14:31004315 - 31153535 on Build GRCh38 | Gain | MIR624 STRN3 AP4S1 HECTD1 |
esv3892495 | Chr.14:30985830 - 31116083 on Build GRCh38 | Gain | MIR624 STRN3 AP4S1 HECTD1 |
esv33189 | Chr.14:30331017 - 31442539 on Build GRCh38 | Loss | LOC101927124 MIR624 G2E3 STRN3 HEATR5A AP4S1 LOC100506071 HECTD1 SCFD1 COCH |
nsv1050054 | Chr.14:31032497 - 31264436 on Build GRCh38 | Gain | AP4S1 HECTD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |