Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | AHNAK2 C14orf79 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | AHNAK2 GPR132 CDCA4 C14orf79 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | AHNAK2 GPR132 JAG2 CDCA4 LOC102723354 C14orf79 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | AHNAK2 CDCA4 C14orf79 PLD4 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | AHNAK2 C14orf79 PLD4 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | LOC105378183 SNORA28 PACS2 INF2 BTBD6 LOC101929634 C14orf79 PLD4 BAG5 LOC107984648 TDRD9 AHNAK2 APOPT1 ZBTB42 MIR203A NUDT14 CDCA4 AKT1 KLC1 RD3L C14orf180 MIR4710 CKB LOC105370691 MIR203B ASPG TRMT61A LOC105370708 ZFYVE21 MARK3 LINC00637 CEP170B XRCC3 BRF1 LOC102723354 TMEM179 KIF26A LOC388022 PPP1R13B LOC105370687 MIR6765 GPR132 LINC00638 JAG2 ADSSL1 EIF5 SIVA1 C14orf2 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | AHNAK2 MIR6765 GPR132 JAG2 BTBD6 NUDT14 CDCA4 BRF1 LOC102723354 C14orf79 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | AHNAK2 MIR6765 GPR132 JAG2 NUDT14 CDCA4 BRF1 LOC102723354 C14orf79 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |