Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AHNAK2 | NM_138420.2 | NP_612429.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1438 | Chr.14:104946391 - 104991244 on Build GRCh38 | Deletion | C14orf79 AHNAK2 |
nsv566141 | Chr.14:104938047 - 105089059 on Build GRCh38 | Gain | CDCA4 GPR132 C14orf79 AHNAK2 |
esv3635634 | Chr.14:104965985 - 105147574 on Build GRCh38 | Gain | CDCA4 JAG2 LOC102723354 GPR132 C14orf79 AHNAK2 |
nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion | CDCA4 PLD4 C14orf79 AHNAK2 |
nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain | PLD4 C14orf79 AHNAK2 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | RD3L NUDT14 BRF1 MIR4710 SNORA28 PACS2 AKT1 ZFYVE21 KIF26A LOC107984648 TRMT61A BTBD6 JAG2 LOC105370687 MIR203A LOC102723354 LOC105370691 ZBTB42 LOC101929634 MIR6765 PLD4 AHNAK2 C14orf180 CDCA4 TDRD9 KLC1 TMEM179 CEP170B MIR203B ASPG BAG5 APOPT1 INF2 C14orf79 C14orf2 LOC105370708 PPP1R13B SIVA1 EIF5 LINC00637 LOC388022 LINC00638 CKB XRCC3 LOC105378183 ADSSL1 GPR132 MARK3 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | CDCA4 NUDT14 JAG2 BRF1 LOC102723354 GPR132 BTBD6 MIR6765 C14orf79 AHNAK2 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | CDCA4 NUDT14 JAG2 BRF1 LOC102723354 GPR132 MIR6765 C14orf79 AHNAK2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |