Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_177533.4 | NP_803877.2 | ||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | 6 | 2466 | ||
BC041584.1 | AAH41584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | JAG2 NUDT14 MIR6765 |
nsv566177 | Chr.14:105167384 - 105177639 on Build GRCh38 | Gain | JAG2 NUDT14 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | PACS2 JAG2 CRIP2 TEX22 LOC100507437 MTA1 BRF1 GPR132 LOC102723354 NUDT14 MIR6765 BTBD6 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | PACS2 JAG2 LOC105370697 IGHA2 TMEM121 CRIP2 TEX22 CRIP1 C14orf80 LOC100507437 MTA1 BRF1 NUDT14 IGH MIR6765 BTBD6 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | JAG2 BRF1 GPR132 LOC102723354 NUDT14 MIR6765 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | JAG2 BRF1 NUDT14 MIR6765 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | JAG2 NUDT14 MIR6765 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | INF2 PLD4 BAG5 ASPG EIF5 AKT1 XRCC3 TDRD9 TRMT61A LINC00638 LOC101929634 AHNAK2 LOC107984648 CKB KLC1 MARK3 ADSSL1 LOC102723354 LOC105370691 C14orf180 BTBD6 CEP170B C14orf2 LOC105370687 PACS2 JAG2 LOC388022 APOPT1 MIR203B RD3L LINC00637 TMEM179 CDCA4 ZBTB42 C14orf79 MIR4710 ZFYVE21 SNORA28 PPP1R13B LOC105378183 BRF1 GPR132 LOC105370708 MIR203A NUDT14 MIR6765 KIF26A SIVA1 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | C14orf79 JAG2 AHNAK2 BRF1 GPR132 CDCA4 LOC102723354 NUDT14 MIR6765 BTBD6 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | IGHG3 IGHV4-31 LOC105370697 IGHA2 MIR4539 IGHV3-23 TMEM121 CRIP2 IGHA1 IGHV3-48 TEX22 CRIP1 MIR8071-1 LOC100507437 MTA1 KIAA0125 MIR4537 MIR4538 BTBD6 IGHV4-55 PACS2 JAG2 IGHV3-30 IGHD IGHV3-69-1 IGHM LINC00221 MIR4507 ADAM6 MIR8071-2 IGHG1 IGHV3-41 IGHG4 LINC00226 C14orf80 ELK2AP BRF1 IGHV1-69 IGHV3-71 IGHG2 NUDT14 IGH MIR6765 LOC107987211 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | C14orf79 JAG2 AHNAK2 BRF1 GPR132 CDCA4 LOC102723354 NUDT14 MIR6765 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |