Assay Details
Target Gene Details
Entrez Gene ID: | 2971 |
Gene Name: | general transcription factor IIIA |
Gene Aliases: |
AP2, TFIIIA |
Location: |
Chr.13:27424544-27435709 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF3A | NM_002097.2 | NP_002088.2 | ||
AK057993.1 | ||||
BP254755.1 | ||||
D32257.1 | BAA06988.1 | |||
U14134.1 | AAA21873.1 | |||
U20272.1 | AAA75623.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |