Assay Details
Target Gene Details
Entrez Gene ID: | 101927155 |
Gene Name: | LMO7 antisense RNA 1 |
Gene Aliases: |
UCHL3-AS1 |
Location: |
Chr.13:75604700-75635994 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LMO7-AS1 | NR_120410.1 | |||
NR_120411.1 | ||||
NR_120412.1 | ||||
AI816891.1 | ||||
AW188034.1 | ||||
BF593247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2751152 | Chr.13:75570063 - 75714282 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv521444 | Chr.13:74139302 - 75679218 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)