Assay Details
Target Gene Details
Entrez Gene ID: | 8847 |
Gene Name: | deleted in lymphocytic leukemia 2 (non-protein coding) |
Gene Aliases: |
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS |
Location: |
Chr.13:49982552-50125541 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLEU2 | NR_002612.1 | |||
AF529010.1 | ||||
AY455757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss | INTS6 DLEU1 SERPINE3 TRIM13 INTS6-AS1 FAM124A DLEU1-AS1 MIR15A SPRYD7 GUCY1B2 LINC00371 DLEU2 DLEU7 ST13P4 MIR5693 RNASEH2B-AS1 DLEU7-AS1 MIR16-1 RNASEH2B MIR3613 KCNRG |
nsv1035151 | Chr.13:49939399 - 50593714 on Build GRCh38 | Loss | DLEU1 TRIM13 DLEU1-AS1 MIR15A MIR16-1 DLEU2 MIR3613 ST13P4 KCNRG |