Assay Details
Target Gene Details
Entrez Gene ID: | 646625 |
Gene Name: | ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5-) decarboxylase |
Gene Aliases: |
PRHOXNB |
Location: |
Chr.13:27978106-27988637 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
URAD | NM_001105577.1 | NP_001099047.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |