Assay Details
Target Gene Details
Entrez Gene ID: | 23026 |
Gene Name: | myosin XVI |
Gene Aliases: |
MYAP3, MYR8, Myo16b, NYAP3, PPP1R107 |
Location: |
Chr.13:108495729-109208007 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYO16 | XM_011521062.1 | XP_011519364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2747975 | Chr.13:108256752 - 108796958 on Build GRCh38 | Deletion | MYO16 TNFSF13B |
nsv1139838 | Chr.13:108256721 - 108796820 on Build GRCh38 | Deletion | MYO16 TNFSF13B |
nsv1050169 | Chr.13:108300121 - 108874622 on Build GRCh38 | Gain | MYO16 TNFSF13B |
nsv832716 | Chr.13:108425141 - 108594609 on Build GRCh38 | Loss | MYO16 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |