Assay Details
Target Gene Details
Entrez Gene ID: | 643246 |
Gene Name: | microtubule associated protein 1 light chain 3 beta 2 |
Gene Aliases: |
ATG8G |
Location: |
Chr.12:116559208-116576620 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAP1LC3B2 | NM_001085481.1 | NP_001078950.1 | ||
XM_011538652.2 | XP_011536954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3630848 | Chr.12:116339176 - 116814937 on Build GRCh38 | Gain | MIR4472-2 LINC00173 MAP1LC3B2 RNFT2 C12orf49 |
esv3630852 | Chr.12:116567434 - 116569881 on Build GRCh38 | Loss | MAP1LC3B2 |
esv2675163 | Chr.12:116567311 - 116569965 on Build GRCh38 | Deletion | MAP1LC3B2 |
nsv1040904 | Chr.12:116562092 - 116596139 on Build GRCh38 | Gain | MAP1LC3B2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |