Assay Details
Target Gene Details
Entrez Gene ID: | 100507424 |
Gene Name: | uncharacterized LOC100507424 |
Gene Aliases: |
- |
Location: |
Chr.12:2836816-2859795 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100507424 | NR_038933.1 | |||
BU947711.1 | ||||
BU947982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain | RHNO1 ITFG2 FOXM1 TULP3 LOC100507424 LOC101929469 FKBP4 NRIP2 |
dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss | RHNO1 ITFG2 FOXM1 TULP3 LOC100507424 LOC101929469 FKBP4 LOC283440 NRIP2 |
nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss | RHNO1 ITFG2 FOXM1 TULP3 LOC100507424 LOC101929469 TEAD4 NRIP2 |