Assay Details
Target Gene Details
Entrez Gene ID: | 121512 |
Gene Name: | FYVE, RhoGEF and PH domain containing 4 |
Gene Aliases: |
CMT4H, FRABP, ZFYVE6 |
Location: |
Chr.12:32399523-32646050 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FGD4 | XM_005253304.4 | XP_005253361.1 | ||
XM_005253309.1 | XP_005253366.1 | |||
XM_017018803.1 | XP_016874292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv558189 | Chr.12:32402912 - 32449145 on Build GRCh38 | Gain | FGD4 |
nsv469346 | Chr.12:32378286 - 32434531 on Build GRCh38 | Loss | BICD1 FGD4 |
esv2751096 | Chr.12:32127406 - 32549850 on Build GRCh38 | Gain | BICD1 FGD4 |
esv3580220 | Chr.12:32404645 - 32462728 on Build GRCh38 | Loss | FGD4 |
esv3629077 | Chr.12:32407211 - 32411434 on Build GRCh38 | Loss | FGD4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |