Assay Details
Target Gene Details
Entrez Gene ID: | 283450 |
Gene Name: | HECT domain E3 ubiquitin protein ligase 4 |
Gene Aliases: |
C12orf51, POTAGE |
Location: |
Chr.12:112160188-112382092 on Build GRCh38 |
Assay Gene Location: | Within Intron 70 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HECTD4 | NM_001109662.3 | NP_001103132.3 | ||
AB014514.3 | BAA31589.3 | |||
AF174498.1 | AAF36539.1 | |||
BC021144.1 | AAH21144.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv483114 | Chr.12:112066462 - 112243129 on Build GRCh38 | Gain | HECTD4 NAA25 TRAFD1 MIR6861 |
esv1004944 | Chr.12:112166125 - 112172944 on Build GRCh38 | Deletion | HECTD4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |