Assay Details
Target Gene Details
Entrez Gene ID: | 10645 |
Gene Name: | calcium/calmodulin dependent protein kinase kinase 2 |
Gene Aliases: |
CAMKK, CAMKKB |
Location: |
Chr.12:121237692-121298308 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv560455 | Chr.12:121087788 - 121363295 on Build GRCh38 | Gain | CAMKK2 P2RX7 ANAPC5 P2RX4 |
nsv1054878 | Chr.12:121082077 - 121328424 on Build GRCh38 | Gain | CAMKK2 P2RX7 ANAPC5 P2RX4 |
nsv952494 | Chr.12:121244498 - 121248497 on Build GRCh38 | Deletion | CAMKK2 |
nsv560454 | Chr.12:121019270 - 121307939 on Build GRCh38 | Loss | CAMKK2 OASL P2RX7 P2RX4 |
nsv455731 | Chr.12:121230451 - 121249371 on Build GRCh38 | Loss | CAMKK2 P2RX4 |
nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain | CAMKK2 P2RX7 XLOC_009911 UNC119B OASL HNF1A-AS1 CABP1 SPPL3 ANAPC5 ACADS MIR4700 C12orf43 HNF1A P2RX4 MLEC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |