Assay Details
Target Gene Details
Entrez Gene ID: | 102724050 |
Gene Name: | uncharacterized LOC102724050 |
Gene Aliases: |
- |
Location: |
Chr.12:54353691-54467030 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102724050 | NR_120486.1 | |||
NR_120487.1 | ||||
DA746386.1 | ||||
DA746474.1 | ||||
DA757660.1 |
Target Gene Details
Entrez Gene ID: | 25946 |
Gene Name: | zinc finger protein 385A |
Gene Aliases: |
HZF, RZF, ZFP385, ZNF385 |
Location: |
Chr.12:54369129-54391609 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF385A | NM_001130967.2 | NP_001124439.1 | ||
NM_001130968.2 | NP_001124440.1 | |||
XM_011538168.2 | XP_011536470.1 | |||
XM_011538169.2 | XP_011536471.1 | |||
XM_011538170.1 | XP_011536472.1 | |||
XM_011538171.2 | XP_011536473.1 | |||
AK024404.1 | BAB14910.1 | |||
AK315613.1 | ||||
BC029752.1 | AAH29752.1 | |||
DB116264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv559000 | Chr.12:54343917 - 54396183 on Build GRCh38 | Loss | COPZ1 GPR84 LOC102724050 ZNF385A ITGA5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |