Assay Details
Target Gene Details
Entrez Gene ID: | 2 |
Gene Name: | alpha-2-macroglobulin |
Gene Aliases: |
A2MD, CPAMD5, FWP007, S863-7 |
Location: |
Chr.12:9067708-9115962 on Build GRCh38 |
Assay Gene Location: | Within Intron 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
A2M | NM_000014.4 | NP_000005.2 | ||
XM_006719056.2 | XP_006719119.1 | |||
AB209614.2 | BAD92851.1 | |||
AK307832.1 | ||||
AK312482.1 | ||||
AY591530.1 | AAT02228.1 | |||
BC026246.1 | AAH26246.1 | |||
BC040071.1 | AAH40071.1 | |||
BX647329.1 | ||||
CR749334.1 | CAH18188.1 | |||
M11313.1 | AAA51551.1 | |||
M36501.1 | AAA51552.1 |
Target Gene Details
Entrez Gene ID: | 10219 |
Gene Name: | killer cell lectin like receptor G1 |
Gene Aliases: |
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE |
Location: |
Chr.12:8989079-9255692 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLRG1 | XM_017018683.1 | XP_016874172.1 | ||
XM_017018684.1 | XP_016874173.1 | |||
XM_017018685.1 | XP_016874174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain | KLRB1 LOC374443 LINC00612 MFAP5 A2M CLEC6A CLECL1 A2MP1 LOC100506159 RIMKLB A2M-AS1 LOC105369728 KLRF2 CLEC4D CD69 LINC00987 KLRF1 AICDA CLEC2A DDX12P OVOS KLRG1 LOC105376678 LOC728715 PZP LOC101930452 LOC642846 CLEC2B CLEC12A A2ML1 M6PR LOC400002 CLEC2D LOC101928030 CLEC4E PHC1 LOC105369649 MIR1244-3 |
nsv832327 | Chr.12:8929183 - 9093624 on Build GRCh38 | Loss | A2M-AS1 LINC00612 KLRG1 M6PR A2M PHC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |