Assay Details
Target Gene Details
Entrez Gene ID: | 3 |
Gene Name: | alpha-2-macroglobulin pseudogene 1 |
Gene Aliases: |
A2MP |
Location: |
Chr.12:9228533-9234207 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
A2MP1 | NR_040112.1 | |||
DB301195.1 |
Target Gene Details
Entrez Gene ID: | 10219 |
Gene Name: | killer cell lectin like receptor G1 |
Gene Aliases: |
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE |
Location: |
Chr.12:8989079-9255692 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLRG1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523958 | Chr.12:9150844 - 9396476 on Build GRCh38 | Loss | MIR1244-3 LOC101928030 PZP LOC101930452 LOC642846 LINC00987 A2MP1 KLRG1 LOC105369649 |
nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain | CLEC6A CLEC4D CLEC4E LOC728715 A2M-AS1 MFAP5 RIMKLB A2M CLEC2D LOC105369649 LOC101928030 LOC642846 OVOS KLRF1 DDX12P A2MP1 KLRB1 KLRF2 CLEC2B M6PR CD69 LOC105369728 MIR1244-3 LOC101930452 LINC00987 A2ML1 PHC1 LOC100506159 CLEC2A CLEC12A AICDA KLRG1 LOC374443 LINC00612 PZP CLECL1 LOC400002 LOC105376678 |
nsv527404 | Chr.12:9098005 - 9282648 on Build GRCh38 | Gain | MIR1244-3 PZP LINC00987 A2MP1 A2M KLRG1 |
esv2759878 | Chr.12:9218999 - 9770933 on Build GRCh38 | Gain+Loss | MIR1244-3 LOC101930452 LINC00987 LOC728715 CLEC2D KLRG1 LOC105369649 LOC374443 LOC101928030 LOC642846 OVOS CLECL1 DDX12P A2MP1 KLRB1 CD69 LOC105376678 LOC105369728 |
nsv557355 | Chr.12:9157399 - 9376419 on Build GRCh38 | Gain | MIR1244-3 LOC101928030 PZP LOC101930452 LOC642846 LINC00987 A2MP1 KLRG1 LOC105369649 |