Assay Details
Target Gene Details
Entrez Gene ID: | 84747 |
Gene Name: | unc-119 lipid binding chaperone B |
Gene Aliases: |
POC7B |
Location: |
Chr.12:120710435-120723640 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UNC119B | NM_001080533.2 | NP_001074002.1 | ||
AK126367.1 | ||||
DA324219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain | C12orf43 ANAPC5 MLEC ACADS HNF1A-AS1 CAMKK2 OASL UNC119B XLOC_009911 MIR4700 SPPL3 P2RX4 CABP1 HNF1A P2RX7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |