Assay Details
Target Gene Details
Entrez Gene ID: | 339 |
Gene Name: | apolipoprotein B mRNA editing enzyme catalytic subunit 1 |
Gene Aliases: |
APOBEC-1, BEDP, CDAR1, HEPR |
Location: |
Chr.12:7649400-7670599 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOBEC1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv521241 | Chr.12:7663155 - 7673709 on Build GRCh38 | Loss | APOBEC1 |
dgv1350n100 | Chr.12:7502541 - 7672399 on Build GRCh38 | Loss | CD163 APOBEC1 |
nsv826213 | Chr.12:7578336 - 7678048 on Build GRCh38 | Gain | APOBEC1 |
nsv557273 | Chr.12:7621432 - 7731755 on Build GRCh38 | Loss | DPPA3 GDF3 CLEC4C APOBEC1 |
dgv5e196 | Chr.12:7621924 - 7954221 on Build GRCh38 | Duplication | DPPA3 NANOGNB GDF3 NANOG SLC2A14 SLC2A3 CLEC4C NANOGP1 APOBEC1 |
nsv521149 | Chr.12:7604110 - 7684392 on Build GRCh38 | Loss | APOBEC1 |