Assay Details
Target Gene Details
Entrez Gene ID: | 83714 |
Gene Name: | nuclear receptor interacting protein 2 |
Gene Aliases: |
- |
Location: |
Chr.12:2825348-2835536 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRIP2 | NM_031474.2 | NP_113662.1 | ||
XM_005253797.3 | XP_005253854.1 | |||
XM_005253798.3 | XP_005253855.1 | |||
XM_011521031.2 | XP_011519333.1 | |||
XM_017020000.1 | XP_016875489.1 | |||
XM_017020001.1 | XP_016875490.1 | |||
AK098063.1 | ||||
AK300944.1 | ||||
AL136557.1 | CAB66492.1 | |||
BC036063.1 | AAH36063.1 | |||
BC131562.1 | ||||
DA391605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3628322 | Chr.12:2796908 - 2837171 on Build GRCh38 | Gain | LOC100507424 FKBP4 ITFG2 NRIP2 |
esv3628321 | Chr.12:2796341 - 2902786 on Build GRCh38 | Gain | LOC101929469 LOC100507424 FOXM1 FKBP4 ITFG2 NRIP2 RHNO1 TULP3 |
dgv19n68 | Chr.12:2726070 - 2935438 on Build GRCh38 | Loss | LOC101929469 LOC100507424 LOC283440 FOXM1 FKBP4 ITFG2 NRIP2 RHNO1 TULP3 |
nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss | LOC101929469 LOC100507424 FOXM1 TEAD4 ITFG2 NRIP2 RHNO1 TULP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |