Assay Details
Target Gene Details
Entrez Gene ID: | 10219 |
Gene Name: | killer cell lectin like receptor G1 |
Gene Aliases: |
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE |
Location: |
Chr.12:8989079-9255692 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLRG1 | XM_017018683.1 | XP_016874172.1 | ||
XM_017018684.1 | XP_016874173.1 | |||
XM_017018685.1 | XP_016874174.1 |
Target Gene Details
Entrez Gene ID: | 253128 |
Gene Name: | long intergenic non-protein coding RNA 612 |
Gene Aliases: |
C12orf33 |
Location: |
Chr.12:9055589-9065070 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00612 | NR_034140.1 | |||
AK123808.1 | ||||
BC028195.1 | AAH28195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain | OVOS M6PR LOC105376678 LINC00612 LOC728715 LOC105369728 LOC642846 CLEC4E CD69 CLEC2A CLEC2B KLRB1 A2ML1 CLECL1 LINC00987 RIMKLB AICDA LOC374443 A2M A2MP1 LOC400002 CLEC4D LOC105369649 A2M-AS1 CLEC6A KLRF2 KLRG1 DDX12P LOC100506159 LOC101930452 PZP LOC101928030 CLEC2D CLEC12A MIR1244-3 PHC1 KLRF1 MFAP5 |
nsv832327 | Chr.12:8929183 - 9093624 on Build GRCh38 | Loss | A2M M6PR A2M-AS1 LINC00612 PHC1 KLRG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |