Assay Details
Target Gene Details
Entrez Gene ID: | 54477 |
Gene Name: | pleckstrin homology domain containing A5 |
Gene Aliases: |
PEPP-2, PEPP2 |
Location: |
Chr.12:19129680-19376400 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041486 | Chr.12:19107917 - 19493057 on Build GRCh38 | Gain | PLEKHA5 SRSF11P1 AEBP2 |
esv2422261 | Chr.12:19109497 - 19353437 on Build GRCh38 | Duplication | PLEKHA5 SRSF11P1 |
esv2422206 | Chr.12:18800088 - 19411427 on Build GRCh38 | Duplication | PLEKHA5 SRSF11P1 AEBP2 |
nsv557675 | Chr.12:19146456 - 19316144 on Build GRCh38 | Loss | PLEKHA5 SRSF11P1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |