Assay Details
Target Gene Details
Entrez Gene ID: | 2967 |
Gene Name: | general transcription factor IIH subunit 3 |
Gene Aliases: |
BTF2, P34, TFB4, TFIIH |
Location: |
Chr.12:123633739-123662604 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF2H3 | NM_001271866.1 | NP_001258795.1 | ||
NM_001271867.1 | NP_001258796.1 | |||
NM_001271868.1 | NP_001258797.1 | |||
NM_001516.4 | NP_001507.2 | |||
XM_017019228.1 | XP_016874717.1 | |||
AK298128.1 | ||||
AK313200.1 | ||||
AL558542.3 | ||||
AL600169.1 | ||||
BC065250.1 | AAH65250.1 | |||
BG720598.1 | ||||
DA953586.1 | ||||
DB170244.1 | ||||
DB482432.1 | ||||
DC319942.1 | ||||
HY046873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759918 | Chr.12:123625575 - 123711309 on Build GRCh38 | Gain | GTF2H3 EIF2B1 TCTN2 |
dgv1566n100 | Chr.12:123610211 - 123711309 on Build GRCh38 | Gain | GTF2H3 EIF2B1 TCTN2 DDX55 |
nsv442292 | Chr.12:123630500 - 123711307 on Build GRCh38 | Gain | GTF2H3 EIF2B1 TCTN2 |
dgv1567n100 | Chr.12:123634608 - 123694870 on Build GRCh38 | Gain | GTF2H3 TCTN2 |
esv35148 | Chr.12:123634573 - 123711573 on Build GRCh38 | Gain | GTF2H3 TCTN2 |
dgv309e214 | Chr.12:123628443 - 123712471 on Build GRCh38 | Gain | GTF2H3 EIF2B1 TCTN2 ATP6V0A2 |
nsv1042181 | Chr.12:123552853 - 123668403 on Build GRCh38 | Gain | GTF2H3 EIF2B1 LOC101927415 TMED2 DDX55 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |