Assay Details
Target Gene Details
Entrez Gene ID: | 54733 |
Gene Name: | solute carrier family 35 member F2 |
Gene Aliases: |
HSNOV1 |
Location: |
Chr.11:107790991-107859188 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC35F2 | NM_017515.4 | NP_059985.2 | ||
AK023080.1 | BAB14394.1 | |||
AK128062.1 | BAC87256.1 | |||
AK300582.1 | ||||
BC039195.2 | AAH39195.1 | |||
BC048302.1 | AAH48302.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv414e212 | Chr.11:107777826 - 107801319 on Build GRCh38 | Gain | SLC35F2 |
dgv2174n54 | Chr.11:107750812 - 107800900 on Build GRCh38 | Gain | SLC35F2 |
nsv442244 | Chr.11:107783017 - 107800462 on Build GRCh38 | Gain | SLC35F2 |
nsv1035229 | Chr.11:107783005 - 107846464 on Build GRCh38 | Gain | SLC35F2 |
dgv22e203 | Chr.11:107776206 - 107801319 on Build GRCh38 | Gain | SLC35F2 |
dgv1274n100 | Chr.11:107597955 - 108083284 on Build GRCh38 | Gain | CUL5 SLN RAB39A SLC35F2 ELMOD1 |
nsv1052198 | Chr.11:107639192 - 107813645 on Build GRCh38 | Gain | SLN SLC35F2 ELMOD1 |
esv2421534 | Chr.11:107782355 - 107800875 on Build GRCh38 | Duplication | SLC35F2 |
nsv1036874 | Chr.11:107777579 - 107809102 on Build GRCh38 | Gain | SLC35F2 |
dgv1276n100 | Chr.11:107693548 - 108059223 on Build GRCh38 | Gain | CUL5 SLN RAB39A SLC35F2 |
dgv230e214 | Chr.11:107682939 - 107838250 on Build GRCh38 | Gain | SLN SLC35F2 |
nsv556380 | Chr.11:107773570 - 107800900 on Build GRCh38 | Gain | SLC35F2 |
esv3892099 | Chr.11:107785223 - 107800900 on Build GRCh38 | Gain | SLC35F2 |
dgv2176n54 | Chr.11:107780369 - 107802574 on Build GRCh38 | Gain | SLC35F2 |
esv3627655 | Chr.11:107740978 - 107953197 on Build GRCh38 | Gain | RAB39A SLC35F2 |
nsv520229 | Chr.11:107778300 - 107799502 on Build GRCh38 | Gain | SLC35F2 |
nsv527748 | Chr.11:107757126 - 107799502 on Build GRCh38 | Gain | SLC35F2 |
nsv521771 | Chr.11:107719609 - 108033920 on Build GRCh38 | Gain | CUL5 RAB39A SLC35F2 |
esv23217 | Chr.11:107782295 - 107801564 on Build GRCh38 | Gain | SLC35F2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |