Assay Details
Target Gene Details
Entrez Gene ID: | 57758 |
Gene Name: | signal peptide, CUB domain and EGF like domain containing 2 |
Gene Aliases: |
CEGB1, CEGF1, CEGP1 |
Location: |
Chr.11:9019500-9091603 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCUBE2 | NM_001170690.1 | NP_001164161.1 | ||
NM_020974.2 | NP_066025.2 | |||
XM_005253032.4 | XP_005253089.2 | |||
XM_005253033.4 | XP_005253090.2 | |||
XM_005253034.4 | XP_005253091.2 | |||
XM_005253035.4 | XP_005253092.2 | |||
XM_005253036.4 | XP_005253093.2 | |||
XM_005253037.4 | XP_005253094.2 | |||
XM_011520246.2 | XP_011518548.1 | |||
XM_011520247.2 | XP_011518549.1 | |||
XM_017018080.1 | XP_016873569.1 | |||
XM_017018081.1 | XP_016873570.1 | |||
XM_017018082.1 | XP_016873571.1 | |||
AK092553.1 | ||||
AK123039.1 | BAC85521.1 | |||
AK131552.1 | BAD18686.1 | |||
BC111690.1 | AAI11691.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33264 | Chr.11:8986065 - 9289584 on Build GRCh38 | Loss | SCUBE2 KRT8P41 DENND5A TMEM41B MIR5691 NRIP3 |
nsv7667 | Chr.11:9016127 - 9050514 on Build GRCh38 | Insertion | SCUBE2 |
esv3891928 | Chr.11:8959990 - 9069697 on Build GRCh38 | Gain | TMEM9B-AS1 SCUBE2 TMEM9B NRIP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |