Assay Details
Target Gene Details
Entrez Gene ID: | 25891 |
Gene Name: | peptidase domain containing associated with muscle regeneration 1 |
Gene Aliases: |
DKFZP586H2123, FP938, RAMP |
Location: |
Chr.11:35431827-35530300 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAMR1 | NM_001001991.2 | NP_001001991.1 | ||
NM_001282675.1 | NP_001269604.1 | |||
NM_001282676.1 | NP_001269605.1 | |||
NM_015430.3 | NP_056245.2 | |||
AF370388.1 | AAQ15224.1 | |||
AK027841.1 | BAB55404.1 | |||
AK297092.1 | ||||
AK300185.1 | ||||
AK316188.1 | ||||
AL832391.1 | CAI46203.1 | |||
AY358346.1 | AAQ88712.1 | |||
BC089434.1 | AAH89434.1 | |||
BX640676.1 | CAE45808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759819 | Chr.11:35297773 - 35458974 on Build GRCh38 | Gain | PAMR1 SLC1A2 |
nsv832120 | Chr.11:35255863 - 35468756 on Build GRCh38 | Gain | PAMR1 SLC1A2 |
nsv1042014 | Chr.11:34919190 - 37195907 on Build GRCh38 | Loss | TRAF6 FJX1 MIR1343 RAG2 RAG1 CD44 MIR3973 COMMD9 C11orf74 PAMR1 TRIM44 LOC100507144 SLC1A2 PDHX PRR5L LOC101928510 LDLRAD3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |