Assay Details
Target Gene Details
Entrez Gene ID: | 283208 |
Gene Name: | prolyl 4-hydroxylase subunit alpha 3 |
Gene Aliases: |
- |
Location: |
Chr.11:74259923-74311676 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P4HA3 | AL833965.1 | CAI46215.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3626835 | Chr.11:74226585 - 74313490 on Build GRCh38 | Gain | PPME1 LOC101928580 P4HA3 |
esv34979 | Chr.11:74167643 - 74822407 on Build GRCh38 | Gain | LOC101928580 P4HA3 LIPT2 C2CD3 CHRDL2 LOC100287896 RNF169 PPME1 POLD3 KCNE3 MIR548AL PGM2L1 MIR4696 |
esv3626833 | Chr.11:74221257 - 74353073 on Build GRCh38 | Gain | PPME1 LOC101928580 P4HA3 PGM2L1 |
esv2759837 | Chr.11:74131564 - 74846552 on Build GRCh38 | Gain | LOC101928580 P4HA3 LIPT2 C2CD3 CHRDL2 LOC100287896 RNF169 PPME1 POLD3 KCNE3 MIR548AL PGM2L1 XRRA1 MIR4696 |
dgv209e214 | Chr.11:74225600 - 74828943 on Build GRCh38 | Gain | RNF169 PPME1 POLD3 KCNE3 LOC101928580 P4HA3 MIR548AL PGM2L1 LIPT2 CHRDL2 MIR4696 LOC100287896 |
nsv825994 | Chr.11:74225622 - 74823410 on Build GRCh38 | Gain | RNF169 PPME1 POLD3 KCNE3 LOC101928580 P4HA3 MIR548AL PGM2L1 LIPT2 CHRDL2 MIR4696 LOC100287896 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |