Assay Details
Target Gene Details
Entrez Gene ID: | 64852 |
Gene Name: | terminal uridylyl transferase 1, U6 snRNA-specific |
Gene Aliases: |
PAPD2, RBM21, STARPAP, TUTase, URLC6 |
Location: |
Chr.11:62575045-62591637 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TUT1 | NM_022830.2 | NP_073741.2 | ||
AB101208.1 | ||||
AK026000.1 | BAB15314.1 | |||
AK292610.1 | ||||
BC110910.1 | AAI10911.1 | |||
BC128263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain | B3GAT3 EEF1G MIR6747 TUT1 EML3 AHNAK MTA2 GANAB ROM1 |
nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss | EEF1G MIR6747 TUT1 AHNAK MTA2 |
nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain | B3GAT3 EEF1G MIR6747 TUT1 EML3 MTA2 GANAB ROM1 |
nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss | UBXN1 EEF1G MIR6747 METTL12 TUT1 EML3 BSCL2 LBHD1 AHNAK INTS5 GANAB UQCC3 LRRN4CL B3GAT3 HNRNPUL2-BSCL2 SNORA57 MTA2 C11orf98 ROM1 |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | UBXN1 EEF1G MIR6747 METTL12 TUT1 GNG3 EML3 BSCL2 LBHD1 INTS5 GANAB HNRNPUL2 UQCC3 LRRN4CL B3GAT3 TTC9C HNRNPUL2-BSCL2 SNORA57 MTA2 C11orf98 ROM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |