Assay Details
Target Gene Details
Entrez Gene ID: | 105369550 |
Gene Name: | STT3A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:125568108-125592568 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STT3A-AS1 | NR_132372.1 | |||
DB453285.1 | ||||
DB470444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss | PATE3 STT3A PATE2 LOC403312 FEZ1 STT3A-AS1 DDX25 PUS3 LOC338667 PATE1 CDON CHEK1 HYLS1 EI24 ACRV1 LOC105369553 PATE4 |