Assay Details
Target Gene Details
Entrez Gene ID: | 334 |
Gene Name: | amyloid beta precursor like protein 2 |
Gene Aliases: |
APLP-2, APPH, APPL2, CDEBP |
Location: |
Chr.11:130069821-130144811 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APLP2 | NM_001142276.1 | NP_001135748.1 | ||
NM_001142277.1 | NP_001135749.1 | |||
NM_001142278.1 | NP_001135750.1 | |||
NM_001243299.1 | NP_001230228.1 | |||
NM_001642.2 | NP_001633.1 | |||
NR_024515.1 | ||||
NR_024516.1 | ||||
AF168956.1 | AAD47291.1 | |||
AK056883.1 | ||||
AK128162.1 | ||||
AK225283.1 | ||||
AK296765.1 | ||||
AK300378.1 | ||||
AK304736.1 | ||||
AK308248.1 | ||||
BC000373.2 | AAH00373.1 | |||
BC004371.1 | AAH04371.1 | |||
DC336210.1 | ||||
L09209.1 | AAA35526.1 | |||
L19597.1 | AAA35601.1 | |||
L27631.1 | AAC41701.1 | |||
S60099.1 | AAC60589.1 | |||
Z22572.1 | CAA80295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759871 | Chr.11:130099327 - 130472822 on Build GRCh38 | Loss | ADAMTS15 APLP2 ZBTB44 ADAMTS8 LOC646383 ST14 |
nsv832295 | Chr.11:130099212 - 130260193 on Build GRCh38 | Gain | APLP2 ZBTB44 ST14 |
nsv556565 | Chr.11:129996758 - 130180927 on Build GRCh38 | Loss | APLP2 LINC00167 ST14 PRDM10 |
dgv251n67 | Chr.11:130005191 - 130124741 on Build GRCh38 | Gain | APLP2 LINC00167 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |