Assay Details
Target Gene Details
Entrez Gene ID: | 283194 |
Gene Name: | uncharacterized LOC283194 |
Gene Aliases: |
- |
Location: |
Chr.11:58933643-59058452 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC283194 | NR_033853.2 | |||
AK091446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv14n68 | Chr.11:58932624 - 59111366 on Build GRCh38 | Gain | GLYATL1 GLYATL1P1 FAM111B LOC283194 |
dgv139e201 | Chr.11:58845738 - 59113363 on Build GRCh38 | Deletion | LOC100287413 GLYATL1 GLYATL1P1 GLYATL2 FAM111B LOC283194 GLYATL1P2 |
nsv1050013 | Chr.11:58627125 - 59453617 on Build GRCh38 | Gain | GLYATL1P1 GLYAT GLYATL2 GLYATL1P2 OR5A1 LOC100287413 GLYATL1 DTX4 OR5AN1 LOC105369314 LOC101927204 OR5A2 FAM111B FAM111A MPEG1 LOC283194 |
nsv482929 | Chr.11:58932626 - 59094208 on Build GRCh38 | Gain | GLYATL1 GLYATL1P1 LOC283194 |
esv2759830 | Chr.11:58932619 - 59241200 on Build GRCh38 | Gain | GLYATL1 DTX4 GLYATL1P1 LOC101927204 FAM111B FAM111A MPEG1 LOC283194 |
nsv469559 | Chr.11:58963312 - 59120166 on Build GRCh38 | Gain | GLYATL1P1 FAM111B LOC283194 |
nsv1159884 | Chr.11:59005164 - 59043992 on Build GRCh38 | Duplication | LOC283194 |
nsv471671 | Chr.11:58963313 - 59120166 on Build GRCh38 | Gain | GLYATL1P1 FAM111B LOC283194 |