Assay Details
Target Gene Details
Entrez Gene ID: | 7536 |
Gene Name: | splicing factor 1 |
Gene Aliases: |
BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162 |
Location: |
Chr.11:64764604-64779043 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528506 | Chr.11:64577976 - 64933558 on Build GRCh38 | Loss | MIR194-2HG SF1 MEN1 NRXN2 PPP2R5B MIR192 ATG2A MAP4K2 EHD1 MIR6749 SLC22A12 MIR6750 RASGRP2 MIR194-2 PYGM CDC42BPG |
nsv469962 | Chr.11:64759608 - 64854101 on Build GRCh38 | Loss | SF1 MEN1 MAP4K2 EHD1 PYGM CDC42BPG |
dgv225n67 | Chr.11:64770620 - 64783455 on Build GRCh38 | Gain | SF1 |
dgv1965n54 | Chr.11:64566642 - 64806117 on Build GRCh38 | Loss | SF1 SLC22A11 MEN1 NRXN2 MAP4K2 PYGM SLC22A12 RASGRP2 |
dgv153n27 | Chr.11:64709578 - 64817487 on Build GRCh38 | Loss | SF1 MEN1 NRXN2 MAP4K2 PYGM RASGRP2 |
nsv832189 | Chr.11:64719711 - 64852727 on Build GRCh38 | Loss | SF1 MEN1 NRXN2 MAP4K2 EHD1 PYGM CDC42BPG RASGRP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |