Assay Details
Target Gene Details
Entrez Gene ID: | 64077 |
Gene Name: | phospholysine phosphohistidine inorganic pyrophosphate phosphatase |
Gene Aliases: |
HDHD2B |
Location: |
Chr.10:124461772-124614141 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LHPP | NM_001167880.1 | NP_001161352.1 | ||
NM_001318331.1 | NP_001305260.1 | |||
NM_001318332.1 | NP_001305261.1 | |||
NM_022126.3 | NP_071409.3 | |||
XM_005270026.3 | XP_005270083.1 | |||
XM_006717943.1 | XP_006718006.1 | |||
XM_011540058.2 | XP_011538360.1 | |||
XM_017016509.1 | XP_016871998.1 | |||
XM_017016510.1 | XP_016871999.1 | |||
XM_017016511.1 | XP_016872000.1 | |||
AK055532.1 | ||||
AW631352.1 | ||||
BC110344.1 | AAI10345.1 | |||
BC113629.1 | ||||
BC113631.1 | ||||
BM541880.1 | ||||
HY154569.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)