Assay Details
Target Gene Details
Entrez Gene ID: | 11155 |
Gene Name: | LIM domain binding 3 |
Gene Aliases: |
CMD1C, CMH24, CMPD3, CYPHER, LDB3Z1, LDB3Z4, LVNC3, MFM4, ORACLE, PDLIM6, ZASP |
Location: |
Chr.10:86664785-86736072 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1049754 | Chr.10:86604960 - 86699322 on Build GRCh38 | Gain |
![]() ![]() |
nsv517139 | Chr.10:86672571 - 86682683 on Build GRCh38 | Gain+Loss |
![]() |
nsv524269 | Chr.10:86654455 - 86680809 on Build GRCh38 | Loss |
![]() ![]() |
dgv116n27 | Chr.10:86653555 - 86682683 on Build GRCh38 | Loss |
![]() ![]() |
dgv948n100 | Chr.10:86646619 - 86714855 on Build GRCh38 | Gain |
![]() ![]() |
nsv551783 | Chr.10:86630480 - 86680809 on Build GRCh38 | Loss |
![]() ![]() |
nsv470968 | Chr.10:86647213 - 86709830 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)