Assay Details
Target Gene Details
Entrez Gene ID: | 105378311 |
Gene Name: | uncharacterized LOC105378311 |
Gene Aliases: |
- |
Location: |
Chr.10:54486230-54656051 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105378311 | NR_134503.1 |
Target Gene Details
Entrez Gene ID: | 65217 |
Gene Name: | protocadherin related 15 |
Gene Aliases: |
CDHR15, DFNB23, USH1F |
Location: |
Chr.10:53802771-55629182 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PCDH15 | NM_001142763.1 | NP_001136235.1 | ||
NM_001142764.1 | NP_001136236.1 | |||
NM_001142765.1 | NP_001136237.1 | |||
NM_001142766.1 | NP_001136238.1 | |||
NM_001142767.1 | NP_001136239.1 | |||
NM_001142768.1 | NP_001136240.1 | |||
NM_001142769.1 | NP_001136241.1 | |||
NM_001142770.1 | NP_001136242.1 | |||
NM_001142771.1 | NP_001136243.1 | |||
NM_001142772.1 | NP_001136244.1 | |||
NM_001142773.1 | NP_001136245.1 | |||
NM_033056.3 | NP_149045.3 | |||
XM_017016571.1 | XP_016872060.1 | |||
XM_017016572.1 | XP_016872061.1 | |||
XM_017016573.1 | XP_016872062.1 | |||
AK126923.1 | ||||
AL834134.1 | CAD38850.2 | |||
AY029205.1 | AAK31581.1 | |||
AY029237.1 | AAK31804.1 | |||
EU718480.1 | ||||
EU718481.1 | ||||
EU718482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041917 | Chr.10:52741572 - 55682853 on Build GRCh38 | Loss | MTRNR2L5 MIR548F1 PCDH15 LOC105378311 MBL2 |
nsv470942 | Chr.10:54485591 - 54529106 on Build GRCh38 | Gain | PCDH15 LOC105378311 |
nsv470943 | Chr.10:54502520 - 54529106 on Build GRCh38 | Loss | PCDH15 LOC105378311 |
dgv1157n54 | Chr.10:54485907 - 54545055 on Build GRCh38 | Loss | PCDH15 LOC105378311 |
nsv818763 | Chr.10:54479670 - 54513840 on Build GRCh38 | Loss | PCDH15 LOC105378311 |
dgv1158n54 | Chr.10:54487871 - 54526308 on Build GRCh38 | Loss | PCDH15 LOC105378311 |
nsv1052386 | Chr.10:54316230 - 54513716 on Build GRCh38 | Gain | PCDH15 LOC105378311 |
esv2750962 | Chr.10:52972795 - 55555375 on Build GRCh38 | Loss | MIR548F1 PCDH15 LOC105378311 |
dgv1146n54 | Chr.10:53034448 - 54727838 on Build GRCh38 | Loss | MIR548F1 PCDH15 LOC105378311 |
dgv1155n54 | Chr.10:54474589 - 54527341 on Build GRCh38 | Loss | PCDH15 LOC105378311 |
dgv1156n54 | Chr.10:54476971 - 54513792 on Build GRCh38 | Loss | PCDH15 LOC105378311 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |