Assay Details
Target Gene Details
Entrez Gene ID: | 55719 |
Gene Name: | SMC5-SMC6 complex localization factor 2 |
Gene Aliases: |
C10orf6, FAM178A |
Location: |
Chr.10:100910230-100965134 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLF2 | NM_001136123.1 | NP_001129595.1 | ||
NM_018121.3 | NP_060591.3 | |||
XM_005269965.2 | XP_005270022.1 | |||
XM_011539944.2 | XP_011538246.1 | |||
AF460991.1 | AAN84475.1 | |||
AK057741.1 | BAB71557.1 | |||
AK292565.1 | ||||
AL832343.1 | ||||
AL832383.1 | ||||
AL833126.1 | ||||
BC047715.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831965 | Chr.10:100916724 - 101082796 on Build GRCh38 | Loss | LZTS2 PDZD7 SFXN3 MIR608 KAZALD1 MRPL43 SEMA4G C10orf2 SLF2 |
nsv552051 | Chr.10:100904892 - 100984619 on Build GRCh38 | Loss | MIR608 MRPL43 SEMA4G SLF2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |