Assay Details
Target Gene Details
Entrez Gene ID: | 84435 |
Gene Name: | adhesion G protein-coupled receptor A1 |
Gene Aliases: |
GPR123 |
Location: |
Chr.10:133070929-133131675 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRA1 | XM_005252695.1 | XP_005252752.1 | ||
AY181241.1 | AAO27353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759793 | Chr.10:133030918 - 133619181 on Build GRCh38 | Gain+Loss | MTG1 KNDC1 SYCE1 TUBGCP2 MIR202HG ADGRA1-AS1 FUOM ECHS1 SPRN VENTX PAOX ADAM8 MIR202 MIR3944 UTF1 CYP2E1 ADGRA1 SCART1 PRAP1 SPRNP1 ZNF511 CALY |
dgv998n100 | Chr.10:132998894 - 133170978 on Build GRCh38 | Gain | KNDC1 ADGRA1 ADGRA1-AS1 |
esv3625022 | Chr.10:132998655 - 133120915 on Build GRCh38 | Gain | ADGRA1 ADGRA1-AS1 |
nsv467498 | Chr.10:132900981 - 133134139 on Build GRCh38 | Gain | ADGRA1 ADGRA1-AS1 CFAP46 LINC01166 LINC01167 LINC01168 |
esv3625015 | Chr.10:132889012 - 133114687 on Build GRCh38 | Gain | ADGRA1 ADGRA1-AS1 CFAP46 LINC01166 LINC01167 LINC01168 |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain | STK32C TCERG1L LOC107984282 BNIP3 ADGRA1-AS1 NKX6-2 LOC105378568 CFAP46 LINC01166 LINC01167 PPP2R2D DPYSL4 INPP5A PWWP2B LRRC27 ADGRA1 MIR378C C10orf91 JAKMIP3 TCERG1L-AS1 LINC01168 LINC01164 |
nsv528260 | Chr.10:132859348 - 133620799 on Build GRCh38 | Loss | MTG1 KNDC1 SYCE1 MIR202HG LINC01167 SPRN ADAM8 ADGRA1 SPRNP1 LINC01168 CALY TUBGCP2 ADGRA1-AS1 FUOM CFAP46 ECHS1 LINC01166 VENTX PAOX MIR202 MIR3944 UTF1 CYP2E1 SCART1 PRAP1 ZNF511 |
nsv952703 | Chr.10:133037897 - 133143896 on Build GRCh38 | Deletion | ADGRA1 ADGRA1-AS1 |
nsv470979 | Chr.10:133033478 - 133118280 on Build GRCh38 | Loss | ADGRA1 ADGRA1-AS1 |
nsv552526 | Chr.10:132699017 - 133103793 on Build GRCh38 | Gain | INPP5A ADGRA1 ADGRA1-AS1 NKX6-2 CFAP46 LINC01166 LINC01167 LINC01168 |
esv3625027 | Chr.10:133082541 - 133084963 on Build GRCh38 | Loss | ADGRA1 |
nsv552539 | Chr.10:133061918 - 133103793 on Build GRCh38 | Loss | ADGRA1 ADGRA1-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |