Assay Details
Target Gene Details
Entrez Gene ID: | 100533184 |
Gene Name: | ARHGAP19-SLIT1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.10:97153042-97292673 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGAP19-SLIT1 | NR_037909.1 | |||
AY336750.1 | AAR02412.1 |
Target Gene Details
Entrez Gene ID: | 6585 |
Gene Name: | slit guidance ligand 1 |
Gene Aliases: |
MEGF4, SLIL1, SLIT-1, SLIT3 |
Location: |
Chr.10:96998038-97185926 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLIT1 | NM_003061.2 | NP_003052.2 | ||
AB011537.3 | BAA32465.3 | |||
AB017167.1 | BAA35184.1 | |||
AY029183.1 | AAK31796.1 | |||
BC146761.1 | ||||
BC146851.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759777 | Chr.10:97030206 - 97202715 on Build GRCh38 | Loss | SLIT1 ARHGAP19-SLIT1 SLIT1-AS1 |
nsv7506 | Chr.10:97142683 - 97180452 on Build GRCh38 | Deletion | SLIT1 ARHGAP19-SLIT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |