Assay Details
Target Gene Details
Entrez Gene ID: | 84504 |
Gene Name: | NK6 homeobox 2 |
Gene Aliases: |
GTX, NKX6.2, NKX6B |
Location: |
Chr.10:132783179-132786033 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NKX6-2 | XM_017016789.1 | XP_016872278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv552528 | Chr.10:132738227 - 132803137 on Build GRCh38 | Gain |
![]() ![]() |
nsv832033 | Chr.10:132743736 - 132870955 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv183n67 | Chr.10:132765626 - 132808425 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv552526 | Chr.10:132699017 - 133103793 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv825660 | Chr.10:132778084 - 132801878 on Build GRCh38 | Gain |
![]() ![]() |
nsv470977 | Chr.10:132699016 - 132814378 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv952700 | Chr.10:132702797 - 132851596 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv825663 | Chr.10:132782861 - 132789273 on Build GRCh38 | Gain |
![]() ![]() |
esv3577894 | Chr.10:132744696 - 132825790 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1054934 | Chr.10:130334322 - 133158679 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv832034 | Chr.10:132761541 - 132940326 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv509378 | Chr.10:132737773 - 132833305 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
nsv832032 | Chr.10:132648722 - 132851966 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv184n67 | Chr.10:132782425 - 132794098 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)