Assay Details
Target Gene Details
Entrez Gene ID: | 54932 |
Gene Name: | exonuclease 3'-5' domain containing 3 |
Gene Aliases: |
mut-7 |
Location: |
Chr.9:137306894-137423262 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EXD3 | NM_017820.4 | NP_060290.3 | ||
XM_011518810.1 | XP_011517112.1 | |||
XM_011518814.1 | XP_011517116.1 | |||
XM_011518815.1 | XP_011517117.1 | |||
XM_017014861.1 | XP_016870350.1 | |||
XM_017014862.1 | XP_016870351.1 | |||
XM_017014863.1 | XP_016870352.1 | |||
XM_017014864.1 | XP_016870353.1 | |||
XM_017014865.1 | XP_016870354.1 | |||
XM_017014866.1 | XP_016870355.1 | |||
XM_017014867.1 | XP_016870356.1 | |||
AB061798.1 | BAC10986.1 | |||
AK055004.1 | BAB70838.1 | |||
AK094438.1 | BAC04356.1 | |||
BU947354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss | TOR4A NDOR1 CYSRT1 TMEM203 EXD3 ANAPC2 NELFB RNF208 RNF224 STPG3 FAM166A TPRN TUBB4B C9orf173-AS1 SLC34A3 SSNA1 NRARP |
dgv13011n54 | Chr.9:137223996 - 137338927 on Build GRCh38 | Loss | RNF224 STPG3 TOR4A FAM166A TUBB4B C9orf173-AS1 CYSRT1 SLC34A3 EXD3 NRARP NELFB |
esv2759721 | Chr.9:137303332 - 137447299 on Build GRCh38 | Loss | EXD3 ENTPD8 NOXA1 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | FBXW5 LCNL1 CYSRT1 NOXA1 MAMDC4 ABCA2 RABL6 ENTPD2 CCDC183-AS1 TPRN C9orf173-AS1 MAN1B1 CCDC183 NRARP MIR7114 FUT7 PHPT1 NDOR1 C9orf142 TMEM203 MIR4479 CLIC3 MIR3621 MAN1B1-AS1 SLC34A3 SSNA1 TRAF2 TOR4A LOC101930307 UAP1L1 RNF208 PRR31 C9orf139 RNF224 STPG3 FAM166A EDF1 TUBB4B GRIN1 ENTPD8 MIR4292 SAPCD2 PNPLA7 NPDC1 C9orf172 DPP7 EXD3 PTGDS C8G LRRC26 ANAPC2 NELFB LCN12 TMEM210 NSMF |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion | TOR4A NDOR1 CYSRT1 TMEM203 EXD3 LRRC26 ANAPC2 NELFB RNF208 TMEM210 MIR3621 RNF224 STPG3 FAM166A TPRN TUBB4B GRIN1 C9orf173-AS1 SLC34A3 SSNA1 NRARP |
nsv6770 | Chr.9:137316337 - 137347440 on Build GRCh38 | Insertion | EXD3 |
nsv8583 | Chr.9:137303248 - 137322124 on Build GRCh38 | Gain | EXD3 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | TOR4A NDOR1 CYSRT1 TMEM203 EXD3 NOXA1 LRRC26 ANAPC2 NELFB RNF208 TMEM210 MIR3621 RNF224 STPG3 FAM166A TPRN TUBB4B GRIN1 C9orf173-AS1 SLC34A3 ENTPD8 SSNA1 NRARP |
nsv1037407 | Chr.9:137275904 - 137361369 on Build GRCh38 | Gain | TOR4A EXD3 NRARP |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | FBXW5 LCNL1 CYSRT1 MAMDC4 ABCA2 SNHG7 RABL6 ENTPD2 CCDC183-AS1 HSPC324 TPRN C9orf173-AS1 MAN1B1 CCDC183 NRARP LCN8 FUT7 LINC01451 MIR6722 PHPT1 NDOR1 EGFL7 C9orf142 TMEM203 MIR4479 CLIC3 MIR3621 MAN1B1-AS1 SNORA17A TMEM141 SLC34A3 SSNA1 TRAF2 TOR4A LOC101930307 LOC100128593 FAM69B UAP1L1 RNF208 PRR31 C9orf139 RNF224 STPG3 FAM166A EDF1 TUBB4B GRIN1 AGPAT2 MIR4292 SAPCD2 NPDC1 C9orf172 DPP7 EXD3 LCN10 LCN15 PTGDS C8G LRRC26 ANAPC2 NELFB LCN6 LCN12 TMEM210 MIR126 SNORA17B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |