Assay Details
Target Gene Details
Entrez Gene ID: | 29126 |
Gene Name: | CD274 molecule |
Gene Aliases: |
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1 |
Location: |
Chr.9:5450503-5470567 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CD274 | NM_001267706.1 | NP_001254635.1 | ||
NM_001314029.1 | NP_001300958.1 | |||
NM_014143.3 | NP_054862.1 | |||
NR_052005.1 | ||||
AF233516.1 | AAG18508.1 | |||
AK300470.1 | ||||
AK314567.1 | ||||
BC069381.1 | AAH69381.1 | |||
DB160805.1 | ||||
DQ836393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759660 | Chr.9:5288908 - 5454512 on Build GRCh38 | Loss | LOC107987017 PLGRKT RLN2 CD274 RLN1 |
nsv613196 | Chr.9:5345673 - 5514372 on Build GRCh38 | Gain | PDCD1LG2 LOC107987017 PLGRKT CD274 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |